• USA Home
  • C6903 - α-Cobratoxin from Naja naja kaouthia

EMAIL THIS PAGE TO A FRIEND
C6903 Sigma

α-Cobratoxin from Naja naja kaouthia

Synonym: Cobratoxin, Snake toxin from Naja naja kaouthia

Purchase

Description

Quality

Chromatographically purified.

Biochem/physiol Actions

Long chain post-synaptic neurotoxin that binds nicotinic receptors and blocks cholinergic neurotransmission at the neuromuscular junction; binding is irreversible.

Price and Availability


Better Together

Discover your Target

Making Headway
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
QQ4498020

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?