• USA Home
  • C6937 - Culture Vessel with Mixer, Nalgene®
EMAIL THIS PAGE TO A FRIEND
C6937 Sigma
Culture Vessel with Mixer, Nalgene®
Purchase
Properties
mfr. no.   Nalgene
Description
Features and Benefits
A complete bioprocess system for large laboratory-scale cell culture or fermentation. Features:
• clear polycarbonate culture vessel graduated to a working capacity of 12 liters
• four access ports on vessel shoulder, with polypropylene closures
• 1/30-hp variable speed mixer drive (110V/220V, 50/60 Hz) with programmable timer, 35-cm shaft and 10-cm impeller
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?