EMAIL THIS PAGE TO A FRIEND
C6960 Sigma

N-Z-Amine® BT

Synonym: Peptone from casein

Purchase

Description

Application

A technical grade analog of N-Z-Amine B. Most useful in applications where clarity is not required.

Other Notes

Casein enzymatic hydrolysate

Legal Information

N-Z-Amine is a registered trademark of Kerry Group

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?