• USA Home
  • C6979 - Monoclonal Anti-Caspase 5 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
C6979 Sigma

Monoclonal Anti-Caspase 5 antibody produced in mouse

~2 mg/mL, clone CAS5, purified immunoglobulin, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to Caspases,
species reactivity   human
application(s)   indirect ELISA: suitable
  microarray: suitable
  western blot: 5-10 μg/mL using a whole extract of HeLa cells activated with lipopolysaccharide (LPS)
clone   CAS5, monoclonal
concentration   ~2 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG2b
mol wt   antigen mol wt ~42 kDa (doublet)
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... CASP5(838)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 300-311 of human caspase 5 protein.

General description

The gene Caspase 5 (CASP5) is mapped to human chromosome 11q22–23. It is a member of the cysteine-aspartic acid protease (caspase) family. CASP5 is particularly detected in placenta, lung, liver, spleen, small intestine, colon and peripheral blood lymphocytes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Application

Monoclonal Anti-Caspase 5 antibody produced in mouse is suitable for the following applications:
• Indirect ELISA
• Microarray
• Western blotting at a concentration of 5-10μg/mL using a whole extract of HeLa cells activated with lipopolysaccharide (LPS)

Biochem/physiol Actions

Caspase-5 (CASP5) mRNA synthesis, protein expression and catalytic activation are highly regulated in response to various pro-inflammatory stimuli, ATP and ER stress inducers. Caspase-5, in association with Caspase-1 may work in modulating human retinal pigment epithelial (hRPE) inflammatory responses. It may have an important role in the inflammatory response in psoriasis and its inactivation may play a role in the tumorigenesis of microsatellite instability (MSI)-positive cancers. CASP5 is up-regulated in inflammatory bowel disease-associated intestinal inflammation. It is elevated in serum of patients with cervical cancer.

Price and Availability


Antibody Explorer

Explore with Confidence

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany  -
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?