• USA Home
  • C7009 - Cholesteryl 11-eicosenoate

EMAIL THIS PAGE TO A FRIEND
C7009 Sigma

Cholesteryl 11-eicosenoate

~95% (HPLC)

Synonym: 3β-Hydroxy-5-cholestene 3-(11-eicosenoate), 5-Cholesten-3β-ol 3-(11-eicosenoate)

Purchase

Properties

assay   ~95% (HPLC)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?