• USA Home
  • C7024 - Creatinase from Flavobacterium sp.

EMAIL THIS PAGE TO A FRIEND
C7024 Sigma

Creatinase from Flavobacterium sp.

lyophilized powder, 10-20 units/mg protein

Synonym: Creatine Amidinohydrolase

Purchase

Description

Unit Definition

One unit will hydrolyze 1.0 μmole of creatine to urea and sarcosine per min at pH 7.5 at 37°C.

Physical form

Lyophilized powder containing phosphate buffer and lactose

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

C3921 Creatinase from Pseudomonas sp., recombinant, expressed in E. coli, lyophilized powder, 10-15 units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?