EMAIL THIS PAGE TO A FRIEND
C7073 Sigma

Cocaethylene-d5

Synonym: 3-Benzyloxy-8-methyl-8-azabicyclo[3.2.1]octane-2-carboxylic acid (ethyl-5) ester

  • CAS Number 145682-86-6

  • Empirical Formula (Hill Notation) C18H18D5NO4

  • Molecular Weight 322.41

  •  PubChem Substance ID 329775139

Purchase

Properties

InChI Key   NMPOSNRHZIWLLL-UFFJXHMVSA-N
form   neat
drug control   Home Office Schedule 2; stupéfiantregulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1648 3/PG 2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?