EMAIL THIS PAGE TO A FRIEND
C7109 Sigma

Cefpirome

solid

Purchase

Properties

form   solid
mol wt   apparent mol wt 514.6

Description

Application

4th generation cephalosporin antibacterial

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?