• USA Home
  • C7127 - Z-Val-Tyr methyl ester

EMAIL THIS PAGE TO A FRIEND
C7127 Sigma

Z-Val-Tyr methyl ester

  • CAS Number 15149-72-1

  • Empirical Formula (Hill Notation) C23H28N2O6

  • Molecular Weight 428.48

  •  MDL number MFCD00020191

  •  PubChem Substance ID 24892933

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   SWPXBDJUGLEKES-UHFFFAOYSA-N
storage temp.   −20°C

Description

Packaging

1 g in poly bottle

Biochem/physiol Actions

Z-Val-Tyr methyl ester (N-CBZ-Val-Tyr-OMe) is an N-terminal and C-terminal blocked derivative of the antihypertensive dipeptide Val-Try (VY).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?