• USA Home
  • C7148 - Z-Leu β-naphthylamide

EMAIL THIS PAGE TO A FRIEND
C7148 Sigma

Z-Leu β-naphthylamide

  • CAS Number 20998-86-1

  • Empirical Formula (Hill Notation) C24H26N2O3

  • Molecular Weight 390.47

  •  MDL number MFCD00056605

  •  PubChem Substance ID 24892940

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, I - Z, Modified Amino Acids
InChI Key   YHLNWQGRZLOYJV-QFIPXVFZSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?