• USA Home
  • C7149 - Cholesterol Oxidase from Pseudomonas fluorescens

EMAIL THIS PAGE TO A FRIEND
C7149 Sigma

Cholesterol Oxidase from Pseudomonas fluorescens

lyophilized powder, 10-50 units/mg protein

Synonym: Cholesterol: oxygen oxidoreductase

Purchase

Description

Other Notes

Contains sodium cholate and borate.

Unit Definition

One unit will convert 1.0 μmole of cholesterol to 4-cholesten-3-one per min at pH 7.5 at 25 °C. Note: 4-cholesten-3-one may undergo isomerization.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Protocols

Enzymatic Assay of Cholesterol Oxidase

To standardize a procedure for the determination of the enzymatic assay of Cholesterol Oxidase.
Keywords: Enzyme activity, Extinction coefficient, Isomerizations

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?