• USA Home
  • C7245 - CAMK1, active, GST tagged from mouse

EMAIL THIS PAGE TO A FRIEND
C7245 Sigma

CAMK1, active, GST tagged from mouse

PRECISIO® Kinase, recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym: A15051 05, D6Ertd263e

Purchase

Properties

Related Categories Active Kinases, Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes,
recombinant   expressed in E. coli
grade   PRECISIO® Kinase
assay   ≥70% (SDS-PAGE)
form   buffered aqueous glycerol solution
activity   2732-3696 nmol/min·mg
mol wt   mol wt ~70 kDa
shipped in   dry ice
storage temp.   −70°C
Gene Information   mouse ... Camk1(52163)

Description

Biochem/physiol Actions

CAMK1 is a serine/threonine protein kinase that is a member of the multifunctional calcium/calmodulin-dependent protein kinase family. CAMK1 is ubiquitously expressed and phosphorylates a number of proteins including SYN1, SYN2, CREB and CFTR. In addition, Numb family of proteins may also be intracellular targets for CAMK1, and they may also be regulated by phosphorylation-dependent interaction with 14-3-3 protein. CAMK1 also plays an important role in the trafficking of HDAC7 between the cytoplasm and the nucleus. CAMK1 phosphorylates HDAC7 on multiple sites that lead to alteration in localization of HDAC7.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?