• USA Home
  • C7256 - Z-Val-Ala methyl ester

EMAIL THIS PAGE TO A FRIEND
C7256 Sigma

Z-Val-Ala methyl ester

  • CAS Number 4817-92-9

  • Empirical Formula (Hill Notation) C17H24N2O5

  • Molecular Weight 336.38

  •  MDL number MFCD00056261

  •  PubChem Substance ID 24892948

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   VSOOFDMWRZMGLN-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?