• USA Home
  • C7337 - Cytochrome c Peptide Map Control from equine heart

EMAIL THIS PAGE TO A FRIEND
C7337 Sigma

Cytochrome c Peptide Map Control from equine heart

lyophilized powder

Purchase

Description

Analysis Note

Tested by trypsin digestion.

Suitability

Suitable as a control for proteolytic digestions.

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
HA5365000

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of Cytochrome c Species Variants on Discovery® BIO PolyMA-SCX (Ion Exchange)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Cytochrome c Species Variants on Discovery® BIO PolyMA-SCX (Ion Exchange)
Keywords: Chromatography, High performance liquid chromatography, Separation

HPLC Analysis of Cytochrome c Species Variants on Discovery® BIO Wide Pore C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Cytochrome c Species Variants on Discovery® BIO Wide Pore C18
Keywords: Chromatography, High performance liquid chromatography

HPLC Analysis of Proteins on Discovery® BIO PolyMA-SCX (Analyte Set #2)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Proteins on Discovery® BIO PolyMA-SCX (Analyte Set #2)
Keywords: Chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?