• USA Home
  • C7358 - N-Acetyl-Ala-Lys-Lys-7-amido-4-methylcoumarin

EMAIL THIS PAGE TO A FRIEND
C7358 Sigma

N-Acetyl-Ala-Lys-Lys-7-amido-4-methylcoumarin

>98% (TLC), solid

Synonym: Ac-AKK-AMC

  • Empirical Formula (Hill Notation) C27H40N6O6

  • Molecular Weight 544.64

  •  MDL number MFCD03452902

  • Popular Documents:  Datasheet (PDF)  

Purchase

Properties

assay   >98% (TLC)
form   solid
storage temp.   −20°C

Description

Application

Plasmin substrate.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

V3138 D-Val-Leu-Lys 7-amido-4-methylcoumarin, ≥98% (HPLC), lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?