• USA Home
  • C7384 - Cholesteryl linolelaidate

EMAIL THIS PAGE TO A FRIEND
C7384 Sigma

Cholesteryl linolelaidate

Synonym: 3β-Hydroxy-5-cholestene 3-linolelaidate, 5-Cholesten-3β-ol 3-linolelaidate, Cholesteryl 9,12-octadecadienoate

Purchase

Properties

Related Categories Biochemicals and Reagents, Cholesterol and Derivatives, Lipids, Sterol Lipids
InChI Key   NAACPBBQTFFYQB-TVYVBBRWSA-N
storage temp.   −20°C

Description

Packaging

Sealed ampule.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?