• USA Home
  • C7442 - Nalgene® jerrican carboy
EMAIL THIS PAGE TO A FRIEND
C7442 Sigma
Nalgene® jerrican carboy
nominal capacity 20 L (5 gal)
Synonym: carboys, jerricans
Purchase
Properties
Related Categories Carboys & Cans, Jerrican, Jerrican carboy, Labware, Manufacturer Browser,
material   high-density polyethylene
  white polypropylene screw closure
mfr. no.   Nalgene 2240 style
nominal capacity   20 L (5 gal)
size   9 in. (229 mm) × 12 in. (305 mm) × 17 1/2 in. (445 mm)
Description
General description
Screw closure is fastened to the carboy with a strap. Graduations are molded in liters and gallons. Recessed bottom provides handgrip for pouring.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?