EMAIL THIS PAGE TO A FRIEND
C75209 Sigma

Cholesterol

94%

Synonym: 3β-Hydroxy-5-cholestene, 5-Cholesten-3β-ol

  • CAS Number 57-88-5

  • Empirical Formula (Hill Notation) C27H46O

  • Molecular Weight 386.65

  •  Beilstein Registry Number 1915888

  •  EC Number 200-353-2

  •  MDL number MFCD00003646

  •  PubChem Substance ID 329775080

  •  eCl@ss 39023139

Purchase

Properties

Related Categories Asymmetric Synthesis, Biochemicals and Reagents, Chemical Synthesis, Chiral Building Blocks, Cholesterol and Derivatives,
biological source   from animal
InChI Key   HVYWMOMLDIMFJA-DPAQBDIFSA-N
assay   94%
form   powder
optical activity   [α]20/D −39°, c = 2 in chloroform
bp   360 °C(lit.)
mp   147-149 °C(lit.)
solubility   water: soluble0.00003 g/L at 20 °C
density   1.067 g/mL at 25 °C(lit.)

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

Biochem/physiol Actions

Major component of all biological membranes; ~25% of total brain lipid is cholesterol.

Price and Availability


Laboratory Gloves

The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
FZ8400000

Frequently Asked Questions

How does the storage temperature relate to shipping conditions?
The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment. http://www.sigmaaldrich.com/etc/medialib/docs/Sigma-Aldrich/General_Information/shippingandlong-termstoragev3.Par.0001.File.tmp/shippingandlong-termstoragev3.pdf
Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis. These documents are located on the product detail page under Useful Links & Tools. Click on the following link to search for a Certificate of Analysis. Please click the following link to see the details on our Product Dating Information.
How do I get lot-specific information or a Certificate of Analysis?
A Certificate of Analysis is available by lot number and can be obtained through our Advanced Search Option: http://www.sigmaaldrich.com/catalog/AdvancedSearchPage.do
How do I find price and availability?
There are several ways to find pricing and availability for our products.  Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers. 
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS. To access the shipping information for this material, use the link on the product detail page for the product, or search here. 
My question is not addressed here, how can I contact Technical Service for assistance?
Use the option to the right to "Ask a Question" by email of a Technical Service Scientist.
What is the solubility of Product No. C75209, Cholesterol?
We test the solubility of C75209 in chloroform at 1 g/4.5 mL (222 mg/mL).  Please see the following link to our specification sheet for this product:  http://www.sigmaaldrich.com/catalog/DataSheetPage.do?brandKey=SIGMA&symbol=C75209
I want to use Product No. C75209, Cholesterol, in cell culture.  Can I dissolve it in ethanol?
One reference cites preparation of a 20 mg/mL solution of cholesterol in ethanol (Razin and Tully, J. Bacteriol., 102(2), 306-310 (1970)).  However, the authors note that the cholesterol crystallizes out in cold storage, so that the solution must be warmed before use.
I want to use Product No. C75209, Cholesterol, in cell culture.  Can I dissolve cholesterol in DMSO?
One reference cites preparation of a 15 mg/mL solution of cholesterol in DMSO (Tomeczkowski et al., Enzyme and Microbial Technology, 15(10), 849-853 (1993)).  However, precipitation of the cholesterol under cold storage is a concern, and solutions may need to be warmed to redissolve any precipitated cholesterol.
Is Product No. C75209, Cholesterol, sterile?
This product is not sterile. Stock solutions would have to be sterile-filtered.
Is Product No. C75209, Cholesterol, soluble in water?
Cholesterol is essentially insoluble in water, less than 0.5 mg/100 mL.
Can I use Product No. C75209, Cholesterol, as an analytical standard?
We do not market Product No. C75209 as an "analytical standard".  It is for general research, so you are free to use this product in analytical work if you wish.
Is Product No. C75209, Cholesterol, USP grade?
Product No. C75209 is not tested to meet USP standards, or any other pharmacopoeia (Ph Eur, BP, JP).
Can I freeze stock solutions of Product No. C75209, Cholesterol?
We have not tested this ourselves, and precipitation of cholesterol from cold stock solutions is a concern. If you freeze or cool stock solutions of cholesterol, it may be necessary to warm them up again to re-dissolve the material. Stock solutions should be protected from light.
Show more questions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?