• USA Home
  • C7538 - Carboxyethylthioethyl 4-O-α-D-galactopyranosyl-β-D-galactopyranoside−BSA conjugate

EMAIL THIS PAGE TO A FRIEND
C7538 Sigma

Carboxyethylthioethyl 4-O-α-D-galactopyranosyl-β-D-galactopyranoside−BSA conjugate

Synonym: (α-D-Gal-[1→4]-β-D-Gal-1→O-CETE)n-BSA

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates A-C, Disaccharide More...
extent of labeling   15-30 mol disaccharide per mol BSA
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?