• USA Home
  • C7608 - N-Acetyl-Ala-Lys-Lys 4-methoxy-2-naphthylamide

EMAIL THIS PAGE TO A FRIEND
C7608 Sigma

N-Acetyl-Ala-Lys-Lys 4-methoxy-2-naphthylamide

solid

Synonym: Ac-AKK-MNA

  • Empirical Formula (Hill Notation) C28H42N6O5

  • Molecular Weight 542.67

  •  MDL number MFCD03452903

  • Popular Documents:  Datasheet (PDF)  

Purchase

Properties

form   solid
storage temp.   −20°C

Description

Application

Plasmin Substrate

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?