EMAIL THIS PAGE TO A FRIEND
C7779 Sigma

3-Chloropyridine

Purchase

Properties

density   1.19 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN2810 - class 6.1 - PG 2 - EHS - Toxic, liquids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
US6125000
Flash Point(F) 
150.8 °F
Flash Point(C) 
66 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

C70002 3-Chloropyridine, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?