• USA Home
  • C7858 - α-B-Crystallin from bovine eye lens

EMAIL THIS PAGE TO A FRIEND
C7858 Sigma

α-B-Crystallin from bovine eye lens

≥90% (SDS-PAGE)

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Cell Stress, Chaperonins and Heat Shock Proteins, Heat Shock and Related Proteins,
assay   ≥90% (SDS-PAGE)
mol wt   mol wt ~20,037
solubility   H2O: soluble
shipped in   dry ice
storage temp.   −70°C
Gene Information   cow ... CRYAB(281719)

Description

Biochem/physiol Actions

Induced by heat and osmotic shock. Closely related to the small heat shock proteins.

Physical form

Solution in PBS, pH 7.2, containing 0.05% sodium azide

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?