EMAIL THIS PAGE TO A FRIEND
C7860 Sigma
Chambered coverslips
3 wells, L × W × D 9.5 mm × 9.5 mm × 1.0 mm
Synonym: chamber slide, chamber slides, glass coverslips
Purchase
Properties
Related Categories Cell Chamber Slides and Glass Coverslips, Cell Culture Supplies, Labware More...
sterility   sterile
L × W × D   9.5 mm × 9.5 mm × 1.0 mm
Description
General description
Chambered coverglass products consist of removable / reusable non-cytotoxic silicone gaskets secured to 24 mm x 50 mm, number 1.5 German coverglass. They may be placed in sterile culture plates or 100 mm diameter culture dishes.
• The thin flat coverglass is the optimal platform for high resolution microscopy an high-sensitivity, quantitative, fluorescent imaging.
• Leak-proof design eliminates well-to-well cross contamination.
• Gaskets stabilize coverglass and provide convenient handling using conventional microscope slide holders and accessories.
• Gaskets are removable for high resolution imaging and may be re-sterilized and reused.
• Well spacing facilitates liquid handling utilizing multichannel pipettes and robotic dispensers.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?