• USA Home
  • C7891 - α-Casein from bovine milk

EMAIL THIS PAGE TO A FRIEND
C7891 Sigma

α-Casein from bovine milk

suitable for substrate for the assay of plasmin (fibrinolysin), ~60% α-casein basis (balance primarily β-casein, electrophoresis), lyophilized powder

Purchase

Properties

assay   ~60% α-casein basis (balance primarily β-casein, electrophoresis)
form   lyophilized powder
suitability   suitable for substrate for the assay of plasmin (fibrinolysin)
storage temp.   −20°C

Description

Quality

Low protease contamination.

Physical form

Contains 10-20% NaCl

Biochem/physiol Actions

α-Casein can be an immunogen in young children, and anti-a-casein antibodies have strong cross-reactivity with insulin. This effect can explain the uncommon insulin autoimmunity, but it does not appear to progress to Type I diabetes.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?