• USA Home
  • C7892 - Cytochrome c from rat heart

EMAIL THIS PAGE TO A FRIEND
C7892 Sigma

Cytochrome c from rat heart

≥95% based on Mol. Wt. 12,132 basis

Purchase

Properties

Related Categories Cell Biology, Cytochrome c, Cytochromes, Electron Transport and Cellular Respiration, Metabolic Pathways,
assay   ≥95% based on Mol. Wt. 12,132 basis
storage temp.   −20°C
Gene Information   rat ... Cycs(25309)

Description

Biochem/physiol Actions

The ready fluctuation of cytochrome c within the cell between ferrous and ferric states, makes it an efficient biological electron-transporter. It plays a vital role in cellular oxidations in both plants and animals. Generally regarded as a universal catalyst of respiration, it forms the essential electron-bridge between the respirable substrates and oxygen.

Preparation Note

Prepared without using TCA.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
HA5365000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

C3131 Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?