• USA Home
  • C8013 - N6-Carboxymethylnicotinamide adenine dinucleotide lithium salt

EMAIL THIS PAGE TO A FRIEND
C8013 Sigma

N6-Carboxymethylnicotinamide adenine dinucleotide lithium salt

~90%

Synonym: NAD-N6-acetic acid

Purchase

Properties

assay   ~90%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?