EMAIL THIS PAGE TO A FRIEND
C8039 Sigma

1-Cyclohexyluracil

powder

Synonym: N-Cyclohexyluracil

  • CAS Number 712-43-6

  • Empirical Formula (Hill Notation) C10H14N2O2

  • Molecular Weight 194.23

  •  MDL number MFCD00038460

  •  PubChem Substance ID 24893014

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   SYRKUUKGYSRJPB-UHFFFAOYSA-N
form   powder
color   faintly yellow
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?