• USA Home
  • C8044 - Comb bridge for Horizontal Submarine units
EMAIL THIS PAGE TO A FRIEND
C8044 Sigma
Comb bridge for Horizontal Submarine units
For 14 cm wide gels
Purchase
Description
Application
Used to hold comb when pouring gels directly on the gel bed.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?