EMAIL THIS PAGE TO A FRIEND
C8048 Sigma

Cholesterylaniline

99%

Synonym: 5-Cholesten-3β-(N-phenyl)amine, 5-Cholestene-3β-(N-phenyl)amine, N-Phenyl-5-cholesten-3β-amine

  • CAS Number 7510-04-5

  • Empirical Formula (Hill Notation) C33H51N

  • Molecular Weight 461.76

  •  MDL number MFCD00046242

  •  PubChem Substance ID 24893016

Purchase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?