• USA Home
  • C8099 - CHO serum-free mediumwithout phenol red and L-glutamine

EMAIL THIS PAGE TO A FRIEND
C8099 Sigma

CHO serum-free mediumwithout phenol red and L-glutamine

Purchase

Properties

Quality Level   GMP
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?