• USA Home
  • C8205 - Cocaethylene solution

EMAIL THIS PAGE TO A FRIEND
C8205 Sigma

Cocaethylene solution

drug standard, 100 μg/mL in acetonitrile (nominal)

Purchase

Properties

Related Categories Additional Standards, Alkaloids, Analytical Standards, Analytical/Chromatography, Chemical Structure,
grade   drug standard
InChI Key   NMPOSNRHZIWLLL-XUWVNRHRSA-N
assay   ≥98%
drug control   Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschlandregulated under CDSA - not available from Sigma-Aldrich Canada
concentration   100 μg/mL in acetonitrile (nominal)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C
Protocols & Articles

Articles

LC/MS Analysis of Cocaine, Cocaethylene, Oxycodone, and Noroxycodone on Discovery® HS F5

From our library of Articles, Sigma-Aldrich presents LC/MS Analysis of Cocaine, Cocaethylene, Oxycodone, and Noroxycodone on Discovery® HS F5
Keywords: Chromatography, Clinical, High performance liquid chromatography

LC/MS Analysis of Illicit Drugs and Metabolites on Ascentis® Express RP-Amide

From our library of Articles, Sigma-Aldrich presents LC/MS Analysis of Illicit Drugs and Metabolites on Ascentis® Express RP-Amide
Keywords: Chromatography, Forensic, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Separation

UHPLC Analysis of Cocaine and Cocaethylene on Titan™ C18

From our library of Articles, Sigma-Aldrich presents UHPLC Analysis of Cocaine and Cocaethylene on Titan™ C18
Keywords: Chromatography, Mass spectrometry

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

C0932 Cocaethylene, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?