• USA Home
  • C8206 - Anti-Calcium Channel (γ2 Subunit) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
C8206 Sigma

Anti-Calcium Channel (γ2 Subunit) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym: Anti-Calcium Channel Caγ2, Anti-Stargazin

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Calcium Channels, Antibodies to Ion Channels,
species reactivity   rat
application(s)   immunohistochemistry: suitable
  western blot (chemiluminescent): 1:200
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   human ... CACNG2(10369)
mouse ... Cacng2(12300)
rat ... Cacng2(84347)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

peptide corresponding to residues 213-228 of mouse or human Caγ2 (with additional C-terminal cysteine). This epitope is identical in mouse and human.

Physical form

Lyophilized at 0.6 mg/ml from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose, and 0.025% sodium azide

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?