EMAIL THIS PAGE TO A FRIEND
C8263 Sigma

Chloroform-d

Synonym: Deuterochloroform

  • CAS Number 865-49-6

  • Linear Formula CDCl3

  • Molecular Weight 120.38

  •  Beilstein Registry Number 1697633

  •  EC Number 212-742-4

  •  MDL number MFCD00000827

Purchase

Properties

InChI Key   HEDRZPFGACZZDS-MICDWDOJSA-N
isotopic purity   100 atom % D
refractive index   n20/D 1.444(lit.)
bp   60.9 °C(lit.)
mp   −64 °C(lit.)
density   1.500 g/mL at 25 °C(lit.)
mass shift   M+1

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 1888 6.1 / PGIII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?