• USA Home
  • C8295 - Silicone sponge closures
EMAIL THIS PAGE TO A FRIEND
C8295 Sigma
Silicone sponge closures
size 18-25 mm
Purchase
Properties
Related Categories Cell Culture Supplies, Culture Flasks, Plastic, Labware More...
size   18-25 mm
Description
Compatibility
For use with 25, 50 mL Erlenmeyer and 25-125 mL DeLong flasks.
General description
Silicone rubber rings with sealed-in silicone sponge filters.
These silicone sponge closures are ideal when working with aerobic cultures; they permit an efficient exchange of gases. They also cut losses due to evaporation in half in comparison to cotton plugs, thus minimizing changes in pH values. They can be dry heat sterilized repeatedly at 180°C and are easy to clean and maintain (reusable).
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?