• USA Home
  • C8357 - Caspase Combo III, Type 6, 8, 9 and 10 human

EMAIL THIS PAGE TO A FRIEND
C8357 Sigma

Caspase Combo III, Type 6, 8, 9 and 10 human

>90% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder

Synonym: Caspase Group III

Purchase

Properties

Related Categories Apoptosis Enzymes, Application Index, Biochemicals and Reagents, Caspases (general), Cell Signaling Enzymes,
recombinant   expressed in E. coli
assay   >90% (SDS-PAGE)
form   lyophilized powder
solubility   PBS: soluble
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... CASP10(843), CASP6(839), CASP8(841), CASP9(842)

Description

Biochem/physiol Actions

Useful for screening caspase inhibitors, studying enzyme kinetics and regulation, determining target substrates, as well as serving as positive controls in caspase activity assays and Western blot analysis.

Unit Definition

One unit will hydrolyze 1 nmol of the caspase substrate VEID-pNA (for caspase-6), IETD-pNA (for caspase-8,10), and LEHD-pNA (for caspase-9), respectively, per hour at pH 7.2 at 37 °C.

Physical form

Contains 0.052% ammonium sulfate, 0.158% Tris−HCl, 0.76% sodium chloride.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?