• USA Home
  • C8387 - Z-Gly-Gly p-nitrophenyl ester

EMAIL THIS PAGE TO A FRIEND
C8387 Sigma

Z-Gly-Gly p-nitrophenyl ester

  • CAS Number 13574-81-7

  • Empirical Formula (Hill Notation) C18H17N3O7

  • Molecular Weight 387.34

  •  MDL number MFCD00038126

  •  PubChem Substance ID 24893056

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   VJPZTTLCVFONEM-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?