• USA Home
  • C8406 - Cobra venom anti-complementary protein from Naja naja kaouthia

EMAIL THIS PAGE TO A FRIEND
C8406 Sigma

Cobra venom anti-complementary protein from Naja naja kaouthia

lyophilized powder

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes, Cell Signaling and Neuroscience,
description   Suitable for inhibition of lysis of antibody sensitized sheep erythrocytes when incubated with human complement serum
form   lyophilized powder
composition   Protein, ~35% biuret
storage temp.   −20°C

Description

Packaging

Package size based on protein content

Physical form

Lyophilized powder containing sodium chloride and ammonium acetate

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?