• USA Home
  • C8476 - Combinatorial Chemistry: A Practical Approach
EMAIL THIS PAGE TO A FRIEND
C8476 Sigma
Combinatorial Chemistry: A Practical Approach
  •  ISBN-10 0-19-963754-7
  •  ISBN-13 978-0-19-963754-6
Purchase
Properties
Related Categories Books and Software, Drug Discovery Books, Labware More...
publication info   H. Fenniri, ed., Oxford University Press, 2000, 432 pp., soft cover
mfr. no.   Oxford University Press
Description
General description
Being a powerful modern technology, combinatorial chemistry has had implications in many areas of chemistry and biology, and the current approaches to drug, catalyst, receptor, and materials development and discovery are all included in this volume. It contains protocols on solid, liquid, and solution phase synthesis and expedient methods of library screening and evaluation, and explains the use of automation and robotics.
Table of Contents
Synthesis of combinatorial libraries using the portioning-mixing
procedure
One-bead-one-compound combinatorial library method
Synthesis and screening of positional scanning synthetic
combinatorial libraries
High throughput combinatorial synthesis of discrete compounds in multimilligram quantities
Positionally addressable parallel synthesis on membrane supports
Resins and anchors for solid phase organic synthesis
Organic reactions on solid support - an overview
Analytical methods in combinatorial chemistry
Multi-component reactions (mcrs) of isocyanides and their chemical libraries
Multi-step solution phase combinatorial synthesis
Experimental techniques in fluorous synthesis
Dendrimer-supported combinatorial chemistry
Automated solution phase synthesis and its application in
combinatorial chemistry
Combinatorial discovery and optimization of electrocatalysts
Combinatorial library synthesis using polymer-supported catalysts
Combinatorial Approaches to chiral catalysts discovery
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?