• USA Home
  • C8497 - Anti-CHN2 (16-30) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
C8497 Sigma

Anti-CHN2 (16-30) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-β-Chimaerin, Anti-ARHGAP3, Anti-BCH, Anti-RHOGAP3

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to G Proteins, Antibodies to G Proteins and Cyclic Nucleotides,
species reactivity   human
application(s)   western blot: 1:500-1:2,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~54 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... CHN2(1124)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 16-30 of human CHN2

General description

α-chimaerin, a GTPase-activating protein gene have two isoforms, α1- and α2-chimaerin. Both the isoforms contain C1 and Rho-GAP domains. But α2 (β) isoform contains an unique N-terminal SH2 domain whereas it is absent in α1. It has a cysteine-rich phorbol ester binding domain. It is expressed in granule cells, specifically in late stage spermatocytes. It has molecular mass of 30-kDa.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Application

Anti-CHN2 (16-30) antibody produced in rabbit is suitable for western blot at a dilution of 1:500-1:2,000.
Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about the Sigma Life Science and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Biochem/physiol Actions

GTPase-activating protein activity of CHN2 is regulated by binding of phospholipid and diacylglycerol (DAG) induces translocation of the protein from the cytosol to the Golgi apparatus membrane.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?