• USA Home
  • C8550 - Spectrophotometer cuvettes, glass
EMAIL THIS PAGE TO A FRIEND
C8550 Sigma
Spectrophotometer cuvettes, glass
Semi-micro, without stopper
Purchase
Properties
Related Categories Absorption Cells, Analytical Reagents, Analytical/Chromatography, Cuvettes, Labware,
material   glass
limit   320-1,000 nm spectral range
H × W × D   45 mm × 12.5 mm × 12.5 mm
chamber volume   ≥1 mL
pathlength   10 mm
Description
General description
See transmission curve labelled E
Price and Availability
Just Prove it! Spectroquant Prove
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?