• USA Home
  • C8585 - 5β-Cholanic acid 3,7-dione methyl ester

EMAIL THIS PAGE TO A FRIEND
C8585 Sigma

5β-Cholanic acid 3,7-dione methyl ester

≥98%

  • CAS Number 7753-72-2

  • Empirical Formula (Hill Notation) C25H38O4

  • Molecular Weight 402.57

  •  MDL number MFCD00271381

Purchase

Properties

Related Categories Bile acids and Derivatives, Biochemicals and Reagents, Lipids, Sterol Lipids
InChI Key   UZRRNRRCPZZPNY-AEVHJRAPSA-N
assay   ≥98%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?