• USA Home
  • C8631 - Cytidine 3′:5′-cyclic mono­phos­phate

EMAIL THIS PAGE TO A FRIEND
C8631 Sigma

Cytidine 3′:5′-cyclic mono­phos­phate

~98%

  • CAS Number 3616-08-8

  • Empirical Formula (Hill Notation) C9H12N3O7P

  • Molecular Weight 305.18

  •  MDL number MFCD00066753

  •  PubChem Substance ID 329775182

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Inhibitors, Enzyme Inhibitors by Type, Enzymes, Inhibitors, and Substrates, Substrate Analogs More...
InChI Key   WCPTXJJVVDAEMW-UHFFFAOYSA-N
assay   ~98%
storage temp.   −20°C

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

C0985 Cytidine 3′,5′-cyclic mono­phos­phate sodium salt, ≥98% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?