• USA Home
  • C8632 - Nα-Carbamyl-L-arginine

EMAIL THIS PAGE TO A FRIEND
C8632 Sigma

Nα-Carbamyl-L-arginine

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   GNRVMKVZOPHHAO-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?