• USA Home
  • C8658 - CM-Cibacron blue 3GA–CM-Agarose

EMAIL THIS PAGE TO A FRIEND
C8658 Sigma

CM-Cibacron blue 3GA–CM-Agarose

saline suspension, Fast flow

Purchase

Properties

Related Categories Cation Exchange Media, Ion Exchange Chromatography, Molecular Biology, Protein Chromatography, Proteomics More...
form   saline suspension
extent of labeling   2-4 μmol per mL
matrix   Cross-linked CM-agarose fast flow
capacity   1,000-2,000 units/mL binding capacity (lactic dehydrogenase (L 2500))(in 0.005 M Tris at pH 8.0 at 20 °C.)
storage temp.   2-8°C

Description

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?