• USA Home
  • C8666 - Monoclonal Anti-Calbindin-D-28K

EMAIL THIS PAGE TO A FRIEND
C8666 Sigma

Monoclonal Anti-Calbindin-D-28K

clone CL-300, whole antiserum

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, CA1-CAL, Primary Antibodies
antibody form   whole antiserum
clone   CL-300, monoclonal
conjugate   unconjugated
shipped in   dry ice
storage temp.   −20°C

Description

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C9848 Monoclonal Anti-Calbindin-D-28K antibody produced in mouse, clone CB-955, ascites fluid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?