• USA Home
  • C8979 - Monoclonal Anti-Cytohesin-1 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
C8979 Sigma

Monoclonal Anti-Cytohesin-1 antibody produced in mouse

~2 mg/mL, clone CYT1-82, purified immunoglobulin, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to ARF and Related Proteins, Antibodies to G Proteins,
species reactivity   human
application(s)   immunocytochemistry: suitable
  indirect ELISA: suitable
  microarray: suitable
  western blot: 2-4 μg/mL using a whole extract of cultured human lymphoma Raji cells
clone   CYT1-82, monoclonal
concentration   ~2 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1
mol wt   antigen mol wt ~50 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... CYTH1(9267)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

recombinant human cytohesin-1.

General description

Cytohesin-1 is a 47kDa protein of B2-1 gene. It is highly expressed in immune cells and is predominantly found in natural killer cells and peripheral T lymphocytes. Its expression has also been reported in purified monocytes and several cultured cell lines. Cytohesin-1 contains two structural motifs: a large central Sec7 domain (200 amino acids) and a carboxy-terminal PH domain (100 amino acids).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Application

Monoclonal Cytohesin-1 antibody is suitable for:
• Western blot at a cocentration of 2-4μg/mL using a whole extract of cultured human lymphoma Raji cells
• Immunocytochemistry
• Indirect ELISA
• Microarray

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Biochem/physiol Actions

Cytohesin-1 is an important factor in vesicular protein trafficking and coatamer assembly in the cell. It also promotes cellular adhesion via binding to the β2 integrin cytoplasmic domain.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  -
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?