EMAIL THIS PAGE TO A FRIEND
C8983 Sigma
Cube Rack
yellow
Purchase
Properties
Related Categories Centrifuge Tube Racks, Labware, Racks More...
material   polypropylene
description   Autoclavable
mfr. no.   Heathrow Scientific
color   yellow
Description
General description
Interlocking on all sides! Each one accommodates (4) 50 mL tubes, (10) 15 mL conical tubes, (12) 12 × 75 or 12 × 100 tubes, or (16) 1.5-2.0 mL microcentrifuge tubes. They slide together on any side allowing the user to work with different tubes on as many connected racks as desired.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?