• USA Home
  • C9015 - Cholesteryl β-D-glucuronide

EMAIL THIS PAGE TO A FRIEND
C9015 Sigma

Cholesteryl β-D-glucuronide

Synonym: 5-Cholesten-3β-ol glucuronide

Purchase

Properties

storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?