EMAIL THIS PAGE TO A FRIEND
C9038 Sigma

Cyclo(Leu-Ala)

Synonym: 3-Isobutyl-6-methyl-2,5-piperazinedione

  • CAS Number 1803-60-7

  • Empirical Formula (Hill Notation) C9H16N2O2

  • Molecular Weight 184.24

  •  MDL number MFCD00079086

  •  PubChem Substance ID 24893130

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   DBJPZCJQDRPOME-UHFFFAOYSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?