• USA Home
  • C9109 - Z-Phe-Phe-fluoromethyl ketone

EMAIL THIS PAGE TO A FRIEND
C9109 Sigma

Z-Phe-Phe-fluoromethyl ketone

≥98% (TLC), solid

Synonym: Z-FF-FMK

Purchase

Price and Availability


Flow Cytometry Instruments

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?